MicroCL 21R Microcentrifuge, refrigerated, 120V, with dual row 18 x 2.0/0.5mL rotor - FAQs
MicroCL 21R Microcentrifuge, refrigerated, 120V, with dual row 18 x 2.0/0.5mL rotor - FAQs
View additional product information for MicroCL 21R Microcentrifuge - FAQs (75002552, 75002561, 75002470)
1 product FAQs found
What rotor does the MicroCL 21R Microcentrifuge, Refrigerated (Cat. No. 75002561) come with?
The MicroCL 21R Microcentrifuge (Cat. No. 75002561) comes with a Dual Row 18 x 2.0/0.5 mL rotor with screw-on lid.