6-Place Fixed-Angle Highconic™ Rotor Adapters
Actual product may vary
6-Place Fixed-Angle Highconic™ Rotor Adapters
6-Place Fixed-Angle Highconic™ Rotor Adapters
6-Place Fixed-Angle Highconic™ Rotor Adapters
6-Place Fixed-Angle Highconic™ Rotor Adapters
Thermo Scientific™
6-Place Fixed-Angle Highconic™ Rotor Adapters
Use these rotor adapters to connect with round or conical-bottom centrifuge tubes and microtubes tubes up to 50mL.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTube Type
75003103Conical Falcon
75003102Conical
7600290616 mL Tube
Catalog number 75003103
Price (USD)
139.33
Online exclusive
158.00
Save 18.67
Each
Tube Type:
Conical Falcon
Price (USD)
139.33
Online exclusive
158.00
Save 18.67
Each
Use these 6-Place Fixed-Angle Highconic™ Rotor Adapters to connect with centrifuge tubes up to 50mL. The adapters accommodate round or conical-bottom tubes and microtubes and work with HiGhConic™ fixed angle rotors.
  • The adapters accommodate round- or conical-bottom tubes and microtubes.
  • Rotor adaptors work with HiGhConic™ fixed angle rotors
  • Fits tube sizes up to 50mL
Specifications
For Use With (Equipment)HiGhConic™ fixed angle rotor
Tube Size50 mL
Tube TypeConical Falcon
Capacity50 mL
TypeRotor Adapter
Unit SizeEach