See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
New Products
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
Home
▸
Shop All Products
▸
Centrifuges
▸
Centrifuge Buckets
▸
H-Flex HS4 Spare Bucket (Set/2)
Thermo Scientific™
H-Flex HS4 Spare Bucket (Set/2)
Spare buckets parts for the H-Flex HS4 rotor
Have Questions?
Catalog Number
For Use With (Equipment)
75003338
H-Flex HS4 Rotor - PN 75003330
Catalog number
75003338
Price (USD)
-
Request A Quote
For Use With (Equipment):
H-Flex HS4 Rotor - PN 75003330
Spare bucket for the H-Flex HS4 rotor
Specifications
Description
H-Flex HS4 Spare Bucket
For Use With (Equipment)
H-Flex HS4 Rotor - PN 75003330
Quantity
2 Pk.