18 x 2.0 mL/0.5 mL Dual Row Rotor with Screw-On-Lid for Sorvall Legend Micro, MicroCL, Pico and Fresco Centrifuges
Thermo Scientific™
18 x 2.0 mL/0.5 mL Dual Row Rotor with Screw-On-Lid for Sorvall Legend Micro, MicroCL, Pico and Fresco Centrifuges
Eliminate the need for adapters with this rotor, which supports two tube volumes in a unique configuration with a screw-on lid.
Have Questions?
Catalog NumberCapacity
7500341818 x 2.0/0.5 mL
Catalog number 75003418
Price (USD)
1,260.00
Online exclusive
1,400.00
Save 140.00
Each
Capacity:
18 x 2.0/0.5 mL
Price (USD)
1,260.00
Online exclusive
1,400.00
Save 140.00
Each
Eliminate the need for adapters with the Thermo Scientific™ 18 x 2.0/0.5mL Dual Row Rotor with Screw-On-Lid, which supports two tube volumes in a unique configuration.
Ordering Information:
Replacement lids sold separately; order rotor body (75003418) and components to complete assembly. For replacement lids order 75003406. Contact your Sales Representative for information.
Compatible with:
Thermo Scientific™ MicroCL 21/21R Microcentrifuge Series.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Angle45°
DescriptionDual Row 18 x 2.0/0.5mL Rotor with Screw-On Lid
For Use With (Equipment)Sorvall Legend Micro 17/17R/21/21R, Pico and Fresco 17/21, MicroCL 17/17R/21/21R Centrifuges
Max. Tube Diameter11 mm
Radius (Metric)8.5 cm
Radius (Metric) Min.4.8 cm
BiocontainmentNo
Capacity18 x 2.0/0.5 mL
MaterialEngineered Polymer
Max. RCF20,800 x g
Max. Speed14,800 rpm
TypeRotor
Unit SizeEach