M-20 Microplate Swinging Bucket Rotor for 1.6 L and 4 L General Purpose X Pro and ST Plus Centrifuges
M-20 Microplate Swinging Bucket Rotor for 1.6 L and 4 L General Purpose X Pro and ST Plus Centrifuges
M-20 Microplate Swinging Bucket Rotor for 1.6 L and 4 L General Purpose X Pro and ST Plus Centrifuges
M-20 Microplate Swinging Bucket Rotor for 1.6 L and 4 L General Purpose X Pro and ST Plus Centrifuges
Thermo Scientific™
M-20 Microplate Swinging Bucket Rotor for 1.6 L and 4 L General Purpose X Pro and ST Plus Centrifuges
Spin up to six microplates and, using sealed carriers, ensure certified sealed conditions for all samples with the Thermo Scientific™ M-20 Microplate Swinging-Bucket Rotor.
Have Questions?
Catalog NumberCapacity
750036242 x 3 Standard or 2 x 1 Deepwell Plates
Catalog number 75003624
Price (USD)
2,267.37
Online exclusive
2,550.00
Save 282.63
Each
Capacity:
2 x 3 Standard or 2 x 1 Deepwell Plates
Price (USD)
2,267.37
Online exclusive
2,550.00
Save 282.63
Each
Spin up to six microplates and, using sealed carriers, ensure certified sealed conditions for all samples with the Thermo Scientific™ M-20 Microplate Swinging-Bucket Rotor.
Ordering Information:
Order 75003625 for Sealed Carriers, with trays and lids (set of 2), and 75002011 for Replacement Sealing Lids (set of 2). For Replacement O-rings for lids (set of 4 with grease) order 75002012. Contact your Sales Representative for information.
Includes:
Two buckets, two adapters
Compatible with:
Thermo Scientific Sorvall™ 16/16R, Sorvall Lengend™ X1/X1R, Multifuge™ X1, Megafuge™ 16 Centrifuge Series
Specifications
Angle90°
Certifications/ComplianceBiocontainment certified by CAMR™, HPA, Porton Down, U.K.
DescriptionM-20 Microplate Rotor
For Use With (Equipment)Thermo Scientific Sorvall Legend XT/XF, Sorvall Legend X1, Sorvall ST 40, Sorvall ST 16, Multifuge X3/X3F, Multifuge X1, Megafuge 40, Megafuge 16 Centrifuge Series
Radius (Metric)12.7 cm
BiocontainmentYes
Capacity2 x 3 Standard or 2 x 1 Deepwell Plates
Max. RCF2,272 x g
Max. Speed4,000 rpm
TypeSwinging Bucket Rotor
Unit SizeEach