TX-750 Round Bucket Adapters
Thermo Scientific™
TX-750 Round Bucket Adapters
Add functionality to your rotor with round bucket adapters, which provide support for round-bottom and conical tubes and bottles, microtubes, and blood collection tubes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity
7500363850 mL Conical Tube
7500363915 mL Conical Tube
75003710250 mL Bottle
75003713100 mL Round-bottom Open-top Tube
7500371550 mL Round-bottom Tube
7500371625 mL Universal Conical/Skirted Tube
7500371814 mL Conical Urine Tube
7500371984 x 15 mL Blood Collection Tube
750037235/7 mL Blood Collection Tube
750037245 mL RIA/Round-bottom Tube (w/o cap)
750037325/7 mL Round-bottom Tube (w/o cap) w/Decanting Aid
750037331.5/2 mL Conical Microtube
75003792250 mL Corning Conical Bottle
Catalog number 75003638
Price (MXN)
-
Capacity:
50 mL Conical Tube
Add functionality to your rotor with Thermo Scientific™ TX-750 Round Bucket Adapters, which provide support for round-bottom and conical tubes and bottles, microtubes and blood collection tubes.
Adapters will hold round-bottom and conical tubes and bottles, microtubes and blood collection tubes.
Specifications
For Use With (Equipment)Round-Bottom and Conical Tube and Bottle, Microtube and Blood Collection Tube
Tube Size50 mL
Tube TypeConical
Capacity50 mL Conical Tube
TypeRound Bucket Adapter
Unit SizePack of 4