Search
| 製品番号(カタログ番号) | 容量 |
|---|---|
| 75003672 | 10 mL Blood Collection Tube |
| 75003671 | 5/7 mL Blood Collection Tube |
| 75003673 | 15 mL Conical Tube |
| 75003674 | 40 x 50 mL Conical Tubes |
| 75003675 | 250 mL Nalgene Oak Ridge Bottles |
| 75003691 | 250 mL Corning Conical Bottles |
| 75003692 | 750 mL Flat-bottom Bottle w/Round Adapters |
Combine Thermo Scientific™ BIOLiner™ Rotor Bucket Adapters with the BIOLiner Rotor Bucket for a complete system.
Ordering Information:
Individual adapters and bottle/adapter packages are offered.
Includes:
75-003-692 includes 750mL flat-bottom bottle with all round adapters for the TX-750 round bucket and four 500mL Corning™ conical bottle adapters.