You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
New Products
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
Home
▸
Shop All Products
▸
Centrifuges
▸
Centrifuge Adapters
▸
TX-400 Rotor Adapters
Thermo Scientific™
TX-400 Rotor Adapters
Use these rotor adapters with round buckets, which enable rotor to accept bottles up to 250 mL and tubes up to 100 mL.
Have Questions?
Change view
Catalog Number
Capacity
75003683
50 mL Conical Tube
75003682
15 mL Conical Tube
75003788
175, 200, 225, 250 mL Bottle
3 Options
Catalog number
75003683
Price (USD)
635.45
Online exclusive
711.00
Save 75.55
Pack of 4
Add to cart
Capacity:
50 mL Conical Tube
Price (USD)
635.45
Online exclusive
711.00
Save 75.55
Pack of 4
Add to cart
Use Thermo Scientific™ TX-400 Adapters with round buckets in the TX-400 Rotor.
The adapters enable rotor to accept bottles up to 250mL and tubes to 100mL.
Maximum adapter capacity: 4 × 250mL (4 × 400mL with 400mL polypropylene bio-bottles)
Maximum adapter O.D. × L: 62 × 125mm (80 × 125mm with 400mL bio-bottles)
Specifications
Max. Tube Diameter
29.5 mm
Max. Tube Length
116 mm
No. per Pack
4
Tube Size
50 mL
Tube Type
Conical
Capacity
50 mL Conical Tube
Type
Rotor Adapter
Unit Size
Pack of 4