Adapters for Double Blood Bag Buckets for Thermo Scientific™ HAEMAFlex™ 12/16 Rotors
Actual product may vary
Adapters for Double Blood Bag Buckets for Thermo Scientific™ HAEMAFlex™ 12/16 Rotors
Adapters for Double Blood Bag Buckets for Thermo Scientific™ HAEMAFlex™ 12/16 Rotors
Adapters for Double Blood Bag Buckets for Thermo Scientific™ HAEMAFlex™ 12/16 Rotors
Adapters for Double Blood Bag Buckets for Thermo Scientific™ HAEMAFlex™ 12/16 Rotors
Thermo Scientific™
Adapters for Double Blood Bag Buckets for Thermo Scientific™ HAEMAFlex™ 12/16 Rotors
Select adapters for double quint blood bags or hook adapters for cord blood separations in double blood bag buckets for Thermo Scientific™ HAEMAFlex™ 12/16 rotors.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity
7500386812/16 x 300 mL
7500385112/16 x 500 mL
7500385212/16 x 500 mL
7500385312/16 x 450 mL
7500385512/16 x 300 mL
Catalog number 75003868
Price (USD)
-
Capacity:
12/16 x 300 mL
Thermo Scientific blood banking centrifuges offer the flexibility to select higher capacity rotors or match existing workflows, with a choice of Thermo Scientific™ HAEMAFlex™ rotors and bucket systems.
For high-throughput blood processing needs, the HAEMAFlex 12 swinging bucket rotor provides 12 x 500mL capacity or upgrade your centrifuge capacity to 16 x 500mL blood bags with the HAEMAFlex 16 rotor and the same bucket selections.
Select adapters for double quint blood bags (XXL, XL or M size) or hook adapters for cord blood separations (XXL or M size).
Specifications
DescriptionHook Adapter for Cord Blood Separations
For Use With (Application)Cord Blood Separations
For Use With (Equipment)XXL Size Adapters
No. per PackSet of 2
Capacity12/16 x 300 mL
TypeHook Adapter
Unit SizeEach