CentriPAK™ Accessory for Sorvall BIOS 16 Bioprocessing Centrifuge
CentriPAK™ Accessory for Sorvall BIOS 16 Bioprocessing Centrifuge
CentriPAK™ Accessory for Sorvall BIOS 16 Bioprocessing Centrifuge
Thermo Scientific™
CentriPAK™ Accessory for Sorvall BIOS 16 Bioprocessing Centrifuge
Thermo Scientific™ CentriPAK™ Accessory, designed for use with CentriPAK BioProcess Container (BPC) which is providing a single-use, sterilized, closed system for cell culture separations in the bioprocessing industry.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionMaterialAutoclavable
75003892CentriPAK BPC Hanging RackStainless SteelNot Autoclavable
75003873CentriPAK BPC AdapterPolypropyleneAutoclavable
75003886CentriPAK BPC ClipPolyethyleneNot Autoclavable
75003889CentriPAK BPC Adapter RackStainless SteelNot Autoclavable
Catalog number 75003892
Price (KRW)
-
Description:
CentriPAK BPC Hanging Rack
Material:
Stainless Steel
Autoclavable:
Not Autoclavable
Accessory for Thermo Scientific™ CentriPAK™ BioProcess Container (BPC). CentriPAK BPC is designed for use in Thermo Scientific™ Sorvall™ BIOS 16 Bioprocessing Centrifuge and 6 x 2000 mL or 8 x 2000 mL Swing Bucket Rotor.
Thermo Scientific™ CentriPAK™ BioProcess Container (BPC) enables you to:
  • Reduce cross-contamination risk while separating cell cultures
  • Perform gentle, high-throughput and high-efficiency harvesting through centrifugation
  • Eliminate post-use cleaning steps required with reusable containers
  • Meet rigorous quality control and assurance practices for the manufacture of industry-standard material components and aseptic product contact film
Specifications
AutoclavableNot Autoclavable
DescriptionCentriPAK BPC Hanging Rack
For Use With (Equipment)CentriPAK BioProcess Container
MaterialStainless Steel
No. per Pack1 Ea.
Product LinePolaris
TypeCentriPAK Accessory
Unit SizeEach