1000mL BIOS Swinging Bucket Rotors
Thermo Scientific™
1000mL BIOS Swinging Bucket Rotors
Efficiently and reliably perform your bioprocessing methods with the Thermo Scientific™ 6 x 1000mL or 8 x 1000mL swinging bucket rotors.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacitydimProductTypeIncludes
750039631000 mLRound Bucket (Set of2)
750039616 x 1000 mLRotor, Round Buckets, 1000 mL Bottles
750039818 x 1000 mLRotor, Round Buckets, 1000 mL Bottles
11776Not ApplicableCover for Round Bucket (set of 2)
12995Not ApplicableSeal Kit (Sets of 6)
Catalog number 75003963
Price (USD)
-
Capacity:
1000 mL
Includes:
Round Bucket (Set of2)
The Thermo Scientific™ 1000mL BIOS swinging bucket rotors are designed for use with the Thermo Scientific™ Sorvall™ BIOS 16 bioprocessing centrifuge. These rotors, which are provided complete with buckets and bottles, have been engineered to simplify operation of the centrifuge while maximizing productivity.
Achieve efficient and reliable sample processing with the following features:
  • 8 x 1000mL (8L) capacity at 5,400 max rpm (8,500 x g max RCF) operation;
  • 6 x 1000mL (6L) capacity at 4,600 max rpm (7,117 x g max RCF) operation
  • Thermo Scientific™ Eco-Spin™ windshield provides energy savings up to 64% when compared with non-windshielded rotors
  • Compatible with Thermo Scientific™ Auto-Lid™ and Auto-ID™ enabling instant lid opening, storage and rotor identification that saves time and protects the integrity of your samples
  • 2-year warranty
Upgrade to Thermo Scientific™ Dura-Coat™ buckets with nickel-coating to improve protection against corrosion due to moisture, chemicals or alkaline solutions.
Specifications
For Use With (Equipment)1000 mL Swinging Bucket Rotor Sets
IncludesRound Bucket (Set of2)
Capacity1000 mL
MaterialStainless Steel Rotor Body, Aluminum Windshield and Bucket, Polypropylene Bottle
TypeBucket
Unit SizeEach