Every Lab Package
Every Lab Package
Every Lab Package
Every Lab Package
Thermo Scientific™
Every Lab Package
Bring your molecular work together by using the Every Lab Package.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct LineRefrigerated
75005527Thermo ScientificYes
75002439SorvallNo
75002562SorvallYes
75003285Thermo ScientificNo
Catalog number 75005527
Price (USD)
-
Product Line:
Thermo Scientific
Refrigerated:
Yes
The Everylab Microcentrifuge and Liquid Handling package provides a convenient offering that includes a microcentrifuge, Finnpipettes F2 GLP Kit (includes 4 single channel pipettes, stand, and tips), and a case of 1.5 mL microcentrifuge tubes (5000 tubes).
Covers basic and complex workflows 
  • Our ventilated and refrigerated microcentrifuges reduce spin times by up to 20%, making them ideal for high-end molecular or genomic research applications
  • The Thermo Scientific microcentrifuges include a 24 X 1.5/2.0mL rotor with ClickSeal™ biocontainment lid allows you to run tubes from 1.5 to 2.0mL tube to mini-preps and spin columns 
  • Pipettes cover a range of volumes to match necessary volumes, from 0.2-2µL to 100-1000µL 
  • Snap Cap tubes offer versatility in the lab: Can be frozen or heated in a water bath
Save time with faster sample processing  
  • Lightweight rotors provide acceleration up to 21,100 x g in 12 seconds.
  • Refrigerated models offer cooling from room temperature to set temperature in just 9 minutes. 
  • Color coding of pipettes helps to select the correct pipette volume while working
Ideal for everyday laboratory use 
  • High performance engineered polymer rotors are designed for outstanding chemical resistance to minimize corrosion from aggressive reagents
  • Pipettes are fully autoclavable, minimizing disruption and downtime
  • Finnpipette pipettes are made of polyvinylidene fluoride (PVDF) components that stand up to harsh chemical, UV light and physical damage
Specifications
IncludesEverylab Centrifuge Refrigerated Package - Thermo Scientific Fresco, 120V (including 24 x 1.5/2 mL rotor); F2 Single Channel Pipettes - GLP Kit 2; Snap Cap tubes (1 case 5000 tubes)
Voltage120 V
Capacity24 x 1.5/2 mL
Max. RCF21,100 x g
Max. Speed14,800 rpm
Product LineThermo Scientific
RefrigeratedYes
Unit SizeEach