Hematocrit Fixed-Angle Rotor
Hematocrit Fixed-Angle Rotor
Hematocrit Fixed-Angle Rotor
Thermo Scientific™
Hematocrit Fixed-Angle Rotor
Save time in clinical processing applications with a fixed-angle rotor that provides dedicated blood processing to achieve 24 capillary separations per run.
Have Questions?
Catalog NumberCapacity
7500573324 Capillary Tubes
Catalog number 75005733
Price (USD)
640.00
Online exclusive
1,600.00
Save 960.00
Each
Capacity:
24 Capillary Tubes
Price (USD)
640.00
Online exclusive
1,600.00
Save 960.00
Each
Save time in clinical processing applications with the Thermo Scientific™ Hematocrit Fixed-Angle Rotor, which provides dedicated blood processing to achieve 24 capillary separations per run.
Achieve red blood cell capillary separations.
Result reading chart provided.
Secure, push-button Auto-Lock rotor exchange in as little as 3 seconds delivers:
  • Trouble-free rotor installation and removal
  • No tools are required
  • No complicated rotor changing procedures
  • Clear chamber access for cleaning convenience
  • Quick switch between applications, from conical tubes to blood tubes, from microtubes to microplates, and the flexibility to evolve with the changing needs of your laboratory
Ordering Information:
Includes linear scale. Replacement capillaries (75000923), sealing kit (75000964) and replacement linear scale (75000938) are sold separately. Contact your sales representative for more information.
Specifications
Angle90°
DescriptionHematocrit Fixed-Angle Rotor
For Use With (Equipment)Thermo Scientific Sorvall ST 8, Megafuge 8 and SL 8 Centrifuges
K Factor1559
Radius (Metric)85 mm
Capacity24 Capillary Tubes
Max. RCF17,860 x g
Max. Speed13,300 rpm
TypeFixed Angle Rotor
Unit SizeEach