Sorvall™ LYNX 4000 Superspeed Centrifuge, 380-400V - FAQs
Sorvall™ LYNX 4000 Superspeed Centrifuge, 380-400V - FAQs
View additional product information for Sorvall LYNX 4000 Superspeed Centrifuge - FAQs (75006581)
1 product FAQs found
What rotor does the Sorvall LYNX 4000 Superspeed Centrifuge (Cat. No. 75006581) come with?
The Sorvall LYNX 4000 Superspeed Centrifuge (Cat. No. 75006581) does not come with any specific rotors included. However, it is compatible with a variety of rotors to fit different applications, supporting labware from microplates, to microtubes, to 1 L bottles (https://assets.thermofisher.com/TFS-Assets/LED/Specification-Sheets/Superspeed-Centrifuge-Rotors-RTBA-EN.pdf).