Sorvall X4R Pro General Purpose Pro Centrifuge, Refrigerated, MD, 120 V - FAQs
Sorvall X4R Pro General Purpose Pro Centrifuge, Refrigerated, MD, 120 V - FAQs
View additional product information for Sorvall X4 and X4R Pro Centrifuges - FAQs (75016078, 75009522, 75016053, 75009820, 75016041, 75016052, 75009520, 75016040, 75016062, 75016084, 75009521, 75009620, 75016061, 75016072, 75009922, 75016071, 75016082, 75009505, 75009920, 75009905, 75009506, 75009507, 75009907)
1 product FAQs found
What centrifuges do you offer that are compatible with TaqMan Array Cards?
Some of the centrifuges we provide that are compatible with TaqMan Array Cards are the Sorvall X4 Pro-MD, 120V (Cat. No. 75009506), ventilated centrifuge, and the Sorvall X4R Pro-MD, 120V (Cat. No. 75009521), refrigerated centrifuge.
You can find a full list of compatible centrifuges in the Centrifuge compatibility for TaqMan Array Cards List.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.