Stable DAB - FAQs
Stable DAB - FAQs
View additional product information for Stable DAB - FAQs (750118)
2 product FAQs found
I am getting high DAB background staining. How can I decrease this background in the future?
High DAB background can be decreased by taking the following steps:
1) Make sure that the stringent wash is performed properly.
2) After hybridization, rinse the slides briefly in room temperature SSC buffer, then immerse slides for 5 minutes in SSC at 75°C.
3) Increase the temperature by 1°C per slide for more than 2 slides, but do not exceed 80°C.
4) For any other wash steps , 1X PBS with Tween 20 (0.025%) is recommended. Washing with any other reagents (1X PBS without Tween 20 or distilled water) can lead to unwanted background.
Do I need to add hydrogen peroxide when using Stable DAB? How stable is it?
Stable DAB is a complete reagent containing DAB and hydrogen peroxide already in the correct amounts. It localizes tissue antigens, DNA, and RNA unambiguously and without background for high-performance immunohistochemistry and in situ hybridization. It creates a non-fading, permanent record of the results as an intense red-brown color. It is stable at -20 degrees C (in a frost-free freezer) for up to 1.5 years.