General Purpose Centrifuge Rotor Packages
Thermo Scientific™
General Purpose Centrifuge Rotor Packages
Everything you need for your swing out rotor in one package
Have Questions?
Change viewbuttonViewtableView
Catalog NumberModel
75016105TX-1000 Cell Culture Package
75016103TX-1000 Blood Tube Package
75016104TX-1000 Microplate Package
75016106TX-200 Cell Culture Package
75016107TX-400 Blood Tube Package
75016108TX-400 Cell Culture Package
75016109TX-750 Blood Tube Package
75016110TX-750 Cell Culture Package
75016111TX-200 Blood Tube Package
Catalog number 75016105
Price (USD)
6,642.00
Online exclusive
7,380.00
Save 738.00
Each
Model:
TX-1000 Cell Culture Package
Price (USD)
6,642.00
Online exclusive
7,380.00
Save 738.00
Each
General Purpose Centrifuge Rotor packages with all the pieces you need to start spinning right away, including a rotor cross, bucket, lids and adapters
Specifications
Certifications/ComplianceISO 14971, ISO 13485, ISO 9001
For Use With (Equipment)Purchase of Thermo Scientific General Purpose Centrifuge required for operation
IncludesTX-1000 with Buckets, ClickSeal Biocontainment lids, 15 and 50 mL conical tube adapters
ModelTX-1000 Cell Culture Package
Net WeightSee individual components of package
WarrantySee Manual
BiocontainmentYes
CapacityMax in TX-1000: 96 x 15 mL Conical Tubes, 40 x 50 mL Conical Tubes, 4 x 1000 mL BioBottles (with TX-1000 Rotors)
MaterialSee individual centrifuge, rotor and accessory data
TypeRotor Equipment Package
Unit SizeEach