7000 Series Copolymer Microsphere Suspensions
Thermo Scientific™

7000 Series Copolymer Microsphere Suspensions

Meet the need for particulate materials with a variety of particle sizes and properties with Thermo Scientific™ 7000 Series Copolymer Microsphere Suspensions.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)Quantity
7503A3.2 μm15 mL
7503B3.2 μm100 mL
7503C3.2 μm1000 mL
7505A6 μm15 mL
7505B6 μm100 mL
7505C6 μm1000 mL
7508A7.9 μm15 mL
7508B7.9 μm100 mL
7510A12 μm15 mL
7510B11 μm100 mL
7510C11 μm1000 mL
7516A17 μm15 mL
7516B17 μm100 mL
7516C17 μm1000 mL
7520A19 μm15 mL
7520B19 μm100 mL
7525A25 μm15 mL
7545A
also known as MCG-7545A-EACH
45 μm15 mL
7545B45 μm100 mL
7550A55 μm15 mL
7550B55 μm100 mL
7550C55 μm1000 mL
7575A
also known as MCG-7575A-EACH
71 μm15 mL
7590A90 μm15 mL
7590B90 μm100 mL
7602A
also known as MCG-7602A-EACH
97 μm15 mL
7602B97 μm100 mL
7640A134 μm15 mL
7725A222 μm15 mL
Catalog number 7503A
Price (USD)
-
Diameter (Metric):
3.2 μm
Quantity:
15 mL
Meet the need for particulate materials with a variety of particle sizes and properties with Thermo Scientific™ 7000 Series Copolymer Microsphere Suspensions, useful as model systems for fluid mechanics experiments and as challenge particles for large pore filtration systems.

Suspensions of large copolymer microspheres are composed of polystyrene polymer, cross-linked with 4 to 8% divinylbenzene (DVB). The particles are chemically inert and can be washed with alcohol and vacuum or air dried. The polymer density is 1.05g/cm3 and the index of refraction is 1.59 at 589nm. They are packaged as aqueous suspensions at 10% solids by weight.

Ordering Information:

15mL, 100mL and 1000mL bottle sizes available. 15mL (“A” Part No. bottles), 100mL (“B” bottles) and 1000mL (“C” bottles) are packaged to order; example (Part No. 7503B and 7503C). Please allow 10 working days when ordering these fill volumes.

Includes:

Material Safety Data Sheet (MSDS) and general product handling insert sheet

Recommended for:

High concentration polymer particles > 3μm used in filter evaluation and testing, fluid mechanics research, dispersion studies and many other research and development projects. Not intended for use in instrument calibration or diagnostic reagents because they lack the exacting specifications needed for those applications. Also useful as experimental particles for acoustical and optical analytical systems.

Not intended for use in instrument calibration or diagnostic reagents.

Not intended for use in instrument calibration or diagnostic reagents.
Specifications
AdditiveContains trace amount of surfactant to inhibit agglomeration and promote stability
Concentration10%
DescriptionParticles useful as model systems for fluid mechanics experiments and as challenge particles for large pore filtration systems.
IncludesGeneral product handling insert sheet
Packaging15 mL
Particle Density1.05g/cm3
Quantity15 mL
Refractive Index1.59 at 589nm (25°C)
StabilityProduct stable for a minimum of 24 months
ColorWhite or Colorless
Diameter (Metric)3.2 μm
For Use With (Application)Fluid Mechanics, Large Pore Filtration
MaterialPolystyrene Divinylbenzene (PS-DVB)
Product TypeCopolymer Microsphere Suspensions
Unit SizeEach
Contents & Storage
Unless otherwise stated, refrigerate (2° to 8°C) product when not in use but do not freeze. Store upright and keep bottle tightly sealed. Mix product with gentle inversion by hand or vortex mixer.