MicroTiter™ Luminescence Strip Microplates (Assembled)
Actual product may vary
MicroTiter™ Luminescence Strip Microplates (Assembled)
MicroTiter™ Luminescence Strip Microplates (Assembled)
Thermo Scientific™

MicroTiter™ Luminescence Strip Microplates (Assembled)

Experience outstanding flexibility in assay design with MicroTiter Luminescence Strip Microplates—1 x12 strips can be broken into any number of wells and placed in the frame.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct Type
7561Microlite 1plus Strip
7562Microlite 2plus Strip
Catalog number 7561
Price (USD)
-
Product Type:
Microlite 1plus Strip

Experience outstanding flexibility in assay design with Thermo Scientific™ MicroTiter Luminescence Strip Microplates. These 1 x12 strips that can be broken into any number of wells and placed in the frame. Thermo Scientific products offer reliability and reproducible results, and are backed by industry-leading quality standards and certifications.

  • Flat bottom
  • White polystyrene
  • Total volume: 380μL
  • Configured in breakable 1 x 12 well strips
  • Choice of MicroLite 1+ or MicroLite 2+ surface
  • Performace assayed using a IgG binding assay

Choice of Surface

  • Microlite 1+ – Hydrophobic surface for preferred binding of biomolecules with hydrophobic domains

  • Microlite 2+ – Slightly hydrophilic surface for increased binding of biomolecules with hydrohillic/hydrophobic character

Ordering Information:

100 units per pack; 100 units per case

Specifications
Binding PropertyHydrophobic
Detection MethodChemiluminescence
For Use With (Application)For preferred binding of biomolecules with hydrophobic domains
FormatStrip Plate
Packaging100 Boxes
Plate BlockingNone
Product TypeMicrolite 1plus Strip
Target MoleculeLipids
Volume (Metric) Working380 μL
ColorWhite
MaterialPolystyrene
No. of Wells12
Product LineMicrotiter
Well ShapeFlat Bottom (F-well)
Unit SizeEach