Immuno Standard Modules White and Black
Immuno Standard Modules White and Black
Immuno Standard Modules White and Black
Immuno Standard Modules White and Black
Immuno Standard Modules White and Black
Thermo Scientific™

Immuno Standard Modules White and Black

Conduct fluorescence- and luminescence-based immunoassays and binding assays using these immuno standard modules white and black.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorVolume (Metric) WellSurface Treatment
7566White380 μLHydrophobic
437591White350 μLMaxiSorp
437702White350 μLPolySorp
437915Clear350 μLMaxiSorp
475515Black400 μLMaxiSorp
475523Black400 μLPolySorp
Catalog number 7566
Price (USD)
308.76
Online exclusive
360.33
Save 51.57
Case of 320
Color:
White
Volume (Metric) Well:
380 μL
Surface Treatment:
Hydrophobic
Price (USD)
308.76
Online exclusive
360.33
Save 51.57
Case of 320
Conduct fluorescence- and luminescence-based immunoassays and binding assays using Thermo Scientific™ Immuno Standard Modules White and Black.

Features:

  • Available in 8-, 12- or 16-well strips
  • Framed or unframed for choice of optimal assay
  • Flat-bottom or C-shaped wells
  • Choice of three surface types for optimal binding
  • White format for highest possible signal reflection with low background fluorescence; recommended for epifluorescence
  • Black format to reduce backscatter light, which may be encountered in epifluorescence
  • Transparent or clear format for use in time-resolved fluorescence applications only

Choice of Surface:

  • Thermo Scientific™ PolySorp™ and Microlite™ 1+ are for adsorption of hydrophobic molecules.
  • Microlite 2+ is slightly hydrophilic and binds a diverse range of biomolecules.
  • Thermo Scientific™ MaxiSorp™ is hydrophilic and ideal for antibody sandwich assay.

Specifications
Binding PropertyHydrophobic
Binding TypeAdsorption
BreakapartBreakable Strips
CertifiedNo
Detection MethodChemiluminescence
For Use With (Application)Luminescence
FormatStrip Plate
LidWithout Lid
No. per Case320
No. per Pack320
Packaging320 Pk., 320 Cases
Plate BlockingNone
Product TypeMicroplate Strip
Target MoleculeLipids
Volume (Metric) Well380 μL
Volume (Metric) Working330 μL
Array1 x 12
ColorWhite
MaterialPolystyrene
No. of Wells96
Surface TreatmentHydrophobic
Well ShapeRound, Flat Bottom (F-well)
Unit SizeCase of 320

Frequently asked questions (FAQs)

Are your Thermo Scientific 8-Well Strip Caps for Immuno Standard Modules compatible with all Nunc MicroWell 96-Well Microplates and Nunc Immuno 96-Well Plates?

Our Thermo Scientific 8-Well Strip Caps for Immuno Standard Modules were designed to provide a positive seal for flat and round bottom wells of our Nunc MicroWell 96-Well Microplates and Nunc Immuno 96-Well Plates.
8-Well Strip Caps for Immuno Standard Modules are not compatible with C or V bottom wells of the Nunc Microwell 96-Well Microplates and Nunc Immuno 96-Well Plates.

For Nunc Immuno plates and modules, what are the advantages of one well geometry type over another? Which types should I use for various applications?

The following list describes the geometries of wells available for Nunc Immuno-plates and modules:
- Flat bottom (F): Allows maximum transmission of light. These plates can be read on a monochromatic reader.
- Round bottom (U): This geometry optimizes washing and coating. These plates can be read using a dual wavelength reader.
- "C" bottom (C): This design of the well is a combination of both flat and round bottoms. Basically, it is a flat bottomed well with curved edges at the bottom. These plates also can be read using a monochromatic reader combining optimal reading and washing.
- StarWell: These wells have a modified "C" shape geometry with eight fins strategically placed at the bottom. This feature increases surface area, allowing more molecules to become immobilized which reduces incubation times.