Bovine Tuberculin PPD is used for sensitive and specific BOVIGAM™ stimulation.
Have Questions?
Catalog Number
Quantity
7600060
5 mL
Catalog number 7600060
Price (USD)
-
Quantity:
5 mL
Bovine Tuberculin PPD is used for sensitive and specific BOVIGAM™ stimulation. PPD is added to the blood sample to stimulate blood lymphocytes to produce IFN-γ.