Adapters for Thermo Scientific™ 24 x 1.5/2.0 mL Rotor with ClickSeal Biocontainment Lid
Thermo Scientific™
Adapters for Thermo Scientific™ 24 x 1.5/2.0 mL Rotor with ClickSeal Biocontainment Lid
Increase your flexibility for micro-volume separations using adapters for our standard microcentrifuge rotor.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacityFor Use With (Equipment)
7600325024 x 0.2 mL PCR Tubes24 x 1.5/2.0mL Rotor with ClickSeal Biocontainment Lid
7600325124 x 0.25/0.4 mL Microliter Tubes24 x 1.5/2.0mL Rotor with ClickSeal Biocontainment Lid
7600325224 x 0.5/0.6 mL Microliter Tubes24 x 1.5/2.0mL Rotor with ClickSeal Biocontainment Lid
Catalog number 76003250
Price (USD)
179.10
Online exclusive
199.00
Save 19.90
Pack of 24
Capacity:
24 x 0.2 mL PCR Tubes
For Use With (Equipment):
24 x 1.5/2.0mL Rotor with ClickSeal Biocontainment Lid
Price (USD)
179.10
Online exclusive
199.00
Save 19.90
Pack of 24
Increase your flexibility for micro-volume separations using adapters for our standard microcentrifuge rotor. Select from a number of Microliter Tube Adapters and PCR adapters for the Thermo Scientific™ 24 x 1.5/2.0mL Rotor with ClickSeal Biocontainment Lid.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
For Use With (Equipment)24 x 1.5/2.0mL Rotor with ClickSeal Biocontainment Lid
Tube Size0.2 mL
Tube TypePCR
Capacity24 x 0.2 mL PCR Tubes
TypePCR Tube Adapter
Unit SizePack of 24