Regulatory requirements vary by country; products may not be available in your geographic area.
PrioCHECK™ Porcine Trichinella Ab Strip Kit
Applied Biosystems™

PrioCHECK™ Porcine Trichinella Ab Strip Kit

Note: The name of this product has changed. It was previously called PRIOCHECK TRICHINELLA AB. The PrioCHECK™ Trichinella Antibody ELISARead more
Have Questions?
Catalog NumberQuantity
7610150450 samples, 5 plates
Catalog number 7610150
Price (USD)
-
Quantity:
450 samples, 5 plates
Note: The name of this product has changed. It was previously called PRIOCHECK TRICHINELLA AB.

The PrioCHECK™ Trichinella Antibody ELISA is a highly sensitive and specific ELISA for the detection of antibodies against Trichinella in serum and meat juice samples of pigs. No cross reactivity with commonly found pig parasites has been observed.

Test procedure
The PrioCHECK™ Trichinella Antibody ELISA is a diagnostic kit based on ELISA technology. It follows a four-step protocol consisting of sample preparation, sample incubation, conjugate incubation, and detection. Serum or meat juice samples are incubated in an ELISA plate coated with the Trichinella E/S antigen. Anti-Trichinella antibodies present in the sample bind to the antigen. Plates are then washed and an anti-swine antibody labeled with horse radish peroxidase (HRP) is used to detect antibodies bound to the E/S antigen. Color development using mittels tetramethyl-benzidine (TMB) substrate measured optically at a wavelength of 450 nm shows the presence of antibodies directed against Trichinella spp.
Specifications
Assay SensitivitySee product documentation
Gene ID (Entrez)Non-Gene
Gene SymbolNon-Gene
Immunoassay Kit GradeStrip
Product LinePrioCHECK, Prionics
Protein FamilyOther Proteins
Quantity450 samples, 5 plates
Target Kit NamedTrichinella Antibody
Target Organism ClassTrichinella
Target Species ValidatedPorcine
Immunoassay Kit FormatOther Format
Target SpeciesPorcine
Unit SizeEach
Contents & Storage
5 Plates