Regulatory requirements vary by country; products may not be available in your geographic area.
PrioCHECK™ FMDV NS Antibody ELISA Kit
Applied Biosystems™

PrioCHECK™ FMDV NS Antibody ELISA Kit

The PrioCHECK™ FMDV NS ELISA is an ELISA that detects antibodies against the highly conserved non-structural (NS) protein of the FMD virus. The test can therefore be used for all species.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
7610440450 Samples
7610760900 Samples
Catalog number 7610440
Price (USD)
-
Quantity:
450 Samples

The PrioCHECK™ FMDV NS ELISA is an ELISA that detects antibodies against the highly conserved non-structural (NS) protein of the FMD virus. The test can therefore be used for all species.

Test procedure
The ELISA test plates of the kit contain the FMDV nonstructural protein, 3ABC, captured by the coated anti-3ABC monoclonal antibody (mAb). A second mAb labeled with an enzyme that generates a color signal (the detection antibody) is then added. The reaction between the FMDV-NS antigen and the detection antibody is blocked by antibodies directed against the non-structural proteins present in the sample. The signal is then measured and when no color is formed, the antibodies in the sample have competed for the viral protein coated on the plate. Samples with a percent inhibition (PI) value of ≥ 50% have shown an immune response to the foot and mouth disease virus. With this result, the sample is immuno-reactive (i.e., anti-FMDV antibodies are present).

Specifications
Assay SensitivitySee product documentation
Gene ID (Entrez)Non-Gene
Gene SymbolNon-Gene
Immunoassay Kit GradeStrip
Product LinePrioCHECK, Prionics
Product TypeELISA Kit
Protein FamilyOther Proteins
Quantity450 Samples
Target Kit NamedFMDV NS Antibody
Target Organism ClassFoot and Mouth Disease Virus (FMDV)
Target Species ValidatedAll Sensitive Animals
Immunoassay Kit FormatOther Format
Target SpeciesBovine
Unit SizeEach
Contents & Storage
5 plates