Regulatory requirements vary by country; products may not be available in your geographic area.
PrioCHECK™ Porcine PRRSV Ab Strip Kit
Applied Biosystems™

PrioCHECK™ Porcine PRRSV Ab Strip Kit

Note: The name of this product has changed. It was previously called PRIOCHECK PRRSV AB . The PrioCHECK™ PRRSV AntibodyRead more
Have Questions?
Catalog NumberQuantity
7610880450 samples, 5 plates
Catalog number 7610880
Price (USD)
-
Quantity:
450 samples, 5 plates
Note: The name of this product has changed. It was previously called PRIOCHECK PRRSV AB .

The PrioCHECK™ PRRSV Antibody ELISA is specially designed for the detection of both the American and the European genotypes of the PRRS virus in serum samples. Plates are coated with ORF 7 protein (common to both genotypes) ensuring the detection of antibodies to both virus types with high sensitivity and specificity. The PrioCHECK™ PRRSV Antibody ELISA shows a high agreement with the most frequently used ELISA in the field and is therefore the most powerful alternative on the PRRS test market.

Test procedure
The PrioCHECK™ PRRSV Antibody ELISA is a diagnostic kit based on ELISA technology. It follows a simple four step protocol consisting of sample preparation, sample incubation, conjugate incubation, and detection. Easy handling makes the test convenient and fast: 90 samples can be analyzed within 150 minutes. Serum samples are incubated in an ELISA plate coated with the ORF 7 protein of PRRSV. Anti-PRRSV antibodies present in the sample bind to the antigen. Plates are then washed and an anti-swine antibody labeled with horse radish peroxidase (HRPO) is used to detect antibodies bound to the ORF 7 coated plate. Color development using tetramethyl-benzidine (TMB) substrate measured optically at a wavelength of 450 nm shows the presence of antibodies directed against PRRSV.
For Veterinary Use Only. For In Vitro Use Only.
Specifications
Assay SensitivitySee product documentation
Immunoassay Kit GradeStrip
Product LinePrioCHECK, Prionics
Protein FamilyOther Proteins
Quantity450 samples, 5 plates
Target Kit NamedPRRSV Antibody
Target Organism ClassPRRSV (Porcine Reproductive and Respiratory Syndrome Virus)
Target Species ValidatedPorcine
Immunoassay Kit FormatOther Format
Target SpeciesPorcine
Unit SizeEach
Contents & Storage
5 Plates