Sodium hydroxide, 95.0-100.5%, pellets, NF, FCC, GMP, Macron Fine Chemicals™
Avantor™

Sodium hydroxide, 95.0-100.5%, pellets, NF, FCC, GMP, Macron Fine Chemicals™

This product is a GMP-manufactured chemical that meets both the F.C.C. and N.F. Requirements.
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
12 kg
25 kg
110 lb.
Packaging:
View all
Poly pail
Catalog number 7680-28
Price (USD)
-
Quantity:
25 kg
Packaging:
Poly pail
This product is a GMP-manufactured chemical that meets both the F.C.C. and N.F. Requirements.
This product is a GMP-manufactured chemical that meets both the F.C.C. and N.F. Requirements.
Specifications
Boiling Point1390°C (1013 hPa)
CAS1310-73-2
Chemical Name or MaterialSodium hydroxide
ColorWhite
Melting Point323°C
Molecular FormulaNaOH
PackagingPoly pail
SynonymSodium hydrate, Soda lye, Caustic soda, Lye
GradeFCC/NF
Molecular Weight (g/mol)40
Percent Purity95.0-100.5%
Quantity25 kg
Unit SizeEach