White 96-Well Immuno Plates
White 96-Well Immuno Plates
White 96-Well Immuno Plates
White 96-Well Immuno Plates
White 96-Well Immuno Plates
Thermo Scientific™

White 96-Well Immuno Plates

Conduct quantitative and qualitative immunoassays like ELISA and binding assays with these white, 96-well immuno plates.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberWell ShapeSurface TreatmentVolume (Metric) Working
7705Round, Flat Bottom (F-well)Microfluor 1280 μL
436110Round, Flat Bottom (F-well)MaxiSorp350 μL
436111Round, Flat Bottom (F-well)PolySorp350 μL
6905TS
also known as 6905
RoundMicrofluor 1280 μL
7571Round, Flat Bottom (F-well)Microlite 1+280 μL
7572Round, Flat Bottom (F-well)Microlite 2+280 μL
7905Round, Flat Bottom (F-well)Microfluor 2280 μL
9502887Round, Flat Bottom (F-well)Universal Binding330 μL
Catalog number 7705
Price (USD)
267.66
Online exclusive
312.62
Save 44.96
Case of 50
Well Shape:
Round, Flat Bottom (F-well)
Surface Treatment:
Microfluor 1
Volume (Metric) Working:
280 μL
Price (USD)
267.66
Online exclusive
312.62
Save 44.96
Case of 50

Conduct quantitative and qualitative immunoassays, like ELISA and binding assays, with Thermo Scientific™ White 96-Well Immuno Plates. These plates, which are compatible with common instruments, deliver optimal results, lot-to-lot reliability and well-to-well reproducibility.

Features

  • Plate has SBS footprint — fits standard equipment
  • Available with flat- or round-bottom wells
  • Choice of three surface types for optimal binding
  • Certified binding homogeneity/reproducibility

Choice of Surface:

  • Thermo Scientific PolySorp, Microfluor 1, Microlite 1+ and Universal Binding (UB) for adsorption of hydrophobic molecules
  • Thermo Scientific MaxiSorp is hydrophilic and ideal for antibody sandwich assays
  • Thermo Scientific Microfluor 2 and Microlite 2+ are slightly hydrophilic and bind a diverse range of biomolecules

White:

  • Gives maximum reflection
  • Minimum autofluorescence and autoluminescence

Compatible with:

Lids for Thermo Scientific MicroWell plates, sealing tape and breathable membrane

Ordering Information:

Availability may vary by country.

Specifications
Binding PropertyHydrophobic
Binding TypeAdsorption
Detection MethodChemiluminescence
For Use With (Application)Fluorescence
FormatStandard
No. per Case50
No. per Pack10
Packaging50 Cases
Plate BlockingNone
Product TypeMicroplate
Target MoleculeLipids
Volume (Metric) Well330 μL
Volume (Metric) Working280 μL
Array8 x 12
ColorWhite
MaterialPolystyrene
No. of Wells96
Surface TreatmentMicrofluor 1
Well ShapeRound, Flat Bottom (F-well)
Unit SizeCase of 50

Frequently asked questions (FAQs)

For Clear Flat-Bottom Immuno Nonsterile 96-Well Plates, is the surface treatment solely on the bottom of the well, or is it also applied to the sides?

For energy treated plates, the entire plate surface receives the energy treatment. The MaxiSorp, PolySorp, MediSorp and MultiSorp plates are energy treated plates. For traditionally treated plates, the well surface is treated up to the listed working volume.

Are your Thermo Scientific 8-Well Strip Caps for Immuno Standard Modules compatible with all Nunc MicroWell 96-Well Microplates and Nunc Immuno 96-Well Plates?

Our Thermo Scientific 8-Well Strip Caps for Immuno Standard Modules were designed to provide a positive seal for flat and round bottom wells of our Nunc MicroWell 96-Well Microplates and Nunc Immuno 96-Well Plates.
8-Well Strip Caps for Immuno Standard Modules are not compatible with C or V bottom wells of the Nunc Microwell 96-Well Microplates and Nunc Immuno 96-Well Plates.

For Nunc Immuno plates and modules, what are the advantages of one well geometry type over another? Which types should I use for various applications?

The following list describes the geometries of wells available for Nunc Immuno-plates and modules:
- Flat bottom (F): Allows maximum transmission of light. These plates can be read on a monochromatic reader.
- Round bottom (U): This geometry optimizes washing and coating. These plates can be read using a dual wavelength reader.
- "C" bottom (C): This design of the well is a combination of both flat and round bottoms. Basically, it is a flat bottomed well with curved edges at the bottom. These plates also can be read using a monochromatic reader combining optimal reading and washing.
- StarWell: These wells have a modified "C" shape geometry with eight fins strategically placed at the bottom. This feature increases surface area, allowing more molecules to become immobilized which reduces incubation times.

Should I use an opaque black/white immuno plate, or an "optical-bottom" design?

Opaque plates can only be used on “top reader” instruments, which detect (or, in fluorescence mode, excite and detect) from above the plate. Readers that detect from underneath the plate (or imaging systems) use optical-bottom plates, which have a black or white upper well structure and a transparent (polystyrene film or optical coverglass) bottom.

Find additional tips, troubleshooting help, and resources within our Antibodies and Immunoassays Support Center.

Why would I use a black or white (instead of clear) immuno plate?

Black plates are often used in assays that make use of a fluorescent reporter because the opaque black surface effectively isolates the signal from each well in order to eliminate “crosstalk” or signal bleed. White plates also isolate well-to-well signals somewhat, but also provide a reflective background to direct luminescent output to the reader, and so often serve as a better choice to improve dynamic range from a luminescent reporter.

Find additional tips, troubleshooting help, and resources within our Antibodies and Immunoassays Support Center.