H-12000 Blood Processing Swinging-Bucket Rotor
Thermo Scientific™
H-12000 Blood Processing Swinging-Bucket Rotor
Achieve various capacities with adapters ranging from 300mL to 2,000mL by using the Thermo Scientific™ H-12000 Blood Processing Swinging-Bucket Rotor.
Have Questions?
Catalog NumberCapacity
7705012 x 500 mL
Catalog number 77050
Price (MXN)
-
Capacity:
12 x 500 mL
Achieve various capacities with adapters ranging from 300mL to 2,000mL by using the Thermo Scientific™ H-12000 Blood Processing Swinging-Bucket Rotor.
Compatible with:
Thermo Scientific™ Sorvall™ RC12BP low-speed centrifuges
Specifications
Angle90°
DescriptionH-12000 Blood Processing Swinging-Bucket Rotor
For Use With (Equipment)Thermo Scientific Sorvall RC 12 Series Large Capacity Centrifuges
K Factor8117
Min. RCF3,610 x g
Radius (Metric)297.2 mm
Radius (Metric) Min.146.3 mm
Capacity12 x 500 mL
MaterialStainless Steel Rotor Body, Aluminum Bucket
Max. RCF7,340 x g
Max. Speed4,700 rpm
TypeSwinging Bucket Rotor
Unit SizeEach