Imject™ Ovalbumin (in PBS)
Thermo Scientific™

Imject™ Ovalbumin (in PBS)

Thermo Scientific Imject OVA is purified ovalbumin protein conveniently formulated for preparation of hapten-carrier conjugates.Features of Imject OVA:• Purified from자세히 알아보기
Have Questions?
카탈로그 번호수량
771205 x 20 mg
카탈로그 번호 77120
제품 가격(KRW)
303,000
온라인 행사
Ends: 30-Sep-2026
356,000
할인액 53,000 (15%)
Each
수량:
5 x 20 mg
대량 주문 또는 맞춤형 요청
제품 가격(KRW)
303,000
온라인 행사
Ends: 30-Sep-2026
356,000
할인액 53,000 (15%)
Each
Thermo Scientific Imject OVA is purified ovalbumin protein conveniently formulated for preparation of hapten-carrier conjugates.

Features of Imject OVA:

• Purified from chicken egg whites and lyophilized in phosphate buffered saline
• Single polypeptide protein with molecular mass of 45,000 Daltons
• Contains 20 lysine residues, most of which have primary amines that are capable of reacting with a conjugation reagent
• Contains 14 aspartic acid and 33 glutamic acid residues whose carboxyl groups account for the net negative charge (pI 4.63) of OVA
• Not as immunogenic as keyhole limpet hemocyanin (KLH) but more soluble in DMSO (dimethyl sulfoxide) for applications that require this solvent
• Useful as an irrelevant protein carrier for antibody screening and immunoassays after using KLH as the carrier protein to generate the immune response against the hapten

Because ovalbumin is only weakly immunogenic, its primary use in antibody production workflows is as a secondary (independent) carrier protein to make antigens more amenable to antibody screening procedures. For example, peptide antigens are more easily coated in polystyrene microplates when conjugated to ovalbumin, thereby facilitating the screening of antisera that were produced using immunogen conjugates with KLH or other more immunogenic carrier protein.

Carrier proteins are large, complex molecules capable of stimulating an immune response upon injection. Successful production of antibodies specific to small antigens (i.e., peptides or drug compounds) requires that these haptens be covalently conjugated to a larger, more complex molecule (usually a protein) to make them immunogenic. Carrier proteins are chosen based on immunogenicity, solubility, and whether adequate conjugation with the carrier can be achieved.

Ovalbumin (OVA), the most abundant protein in chicken egg whites, is a convenient protein for a variety of uses in the laboratory. The 45 kDa protein is not as immunogenic as other proteins, such as keyhole limpet hemocyanin (KLH). However, OVA's independent structure make it especially useful as a carrier for peptides and other small antigens that by themselves are difficult to manipulate in typical screening assays such as ELISA. Although it is more hydrophobic and less soluble in aqueous buffers than BSA, ovalbumin remains soluble in up to 70% DMSO, making it useful for conjugation with haptens that require DMSO for solubility. Ovalbumin contains numerous primary amines and carboxyl groups that can be targeted for conjugation with glutaraldehyde, NHS-ester, EDC and other crosslinking reagents.

Related Products
Imject™ Maleimide-Activated Ovalbumin
For Research Use Only. Not for use in diagnostic procedures.
사양
수량5 x 20 mg
제품라인Imject
Unit SizeEach
구성 및 보관
Supplied: Carrier proteins are supplied lyophilized. When reconstituted with 2mL of ultrapure water the buffer compositions for BSA and mcKLH is 0.1M sodium phosphate, 0.15M NaCl; pH 7.2 and for OVA is 0.01M sodium phosphate, pH 7.2. Proteins contain proprietary stabilizers.

Storage: Upon receipt store product at 4°C. Product is shipped at ambient temperature