Imject™ Maleimide-Activated mcKLH
Thermo Scientific™
Imject™ Maleimide-Activated mcKLH
Thermo Scientific Imject Maleimide-Activated mcKLH enables conjugation of sulfhydryl-containing peptide haptens to the highly immunogenic keyhole limpet hemocyanin (KLH) carrierRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
7761010 x 10 mg
7760510 mg
776062 mg
Catalog number 77610
Price (USD)
4,595.00
Each
Quantity:
10 x 10 mg
Request bulk or custom format
Price (USD)
4,595.00
Each
Thermo Scientific Imject Maleimide-Activated mcKLH enables conjugation of sulfhydryl-containing peptide haptens to the highly immunogenic keyhole limpet hemocyanin (KLH) carrier protein for immunization and antibody production.
Features of Imject Maleimide-Activated mcKLH:
• Optimized activation—specifications require that each lot is activated with more than 400 moles of sulhydryl-reactive maleimide per mole of KLH, ensuring consistently high levels of conjugation
• Sulfhydryl-reactive—engineered for conjugating cysteine-containing peptide antigens to make effective immunogens for high-titer antibody production
• Simple, rapid procedure—complete the hapten-carrier conjugation reaction in two hours to yield sufficient immunogen for several injections and booster immunizations of host animals
• High-quality carrier protein—KLH is isolated from the mollusk Megathura crenulata (keyhole limpet); mcKLH is a high-quality form of KLH that is harvested from limpets grown in mariculture
• Highly immunogenic—KLH has a high molecular mass (4.5 x 105 to 1.3 x 107 Daltons; aggregates of 350 and 390 kDa subunits) and elicits a stronger immune response than BSA or ovalbumin
• Excellent solubility—our preparations of mariculture KLH are more soluble than typical forms
The high-quality mariculture KLH is rendered sulfhydryl-reactive by chemical modification with Sulfo-SMCC crosslinker, and the resulting maleimide-activated protein is then stabilized and lyophilized in phosphate buffer. Maleimide groups form covalent crosslinks with sulfhydryl (-SH) moieties on cysteine residues of peptides and other thiol-containing haptens. Numerous antigen molecules can be conjugated to each KLH molecule to make them highly immunogenic and increase the likelihood of eliciting immunized animals to produce high-titer antisera to the epitopes of interest. Select from among several package sizes of activated KLH protein, or choose the complete conjugation kit that includes optimized buffers and trouble-free spin desalting columns.
Carrier proteins are large, complex molecules capable of stimulating an immune response upon injection. Successful production of antibodies specific to small antigens (i.e., peptides or drug compounds) requires that these haptens be covalently conjugated to a larger, more complex molecule (usually a protein) to make them immunogenic. Carrier proteins are chosen based on immunogenicity, solubility, and whether an adequate level of hapten-carrier conjugation can be achieved.
Keyhole limpet hemocyanin (KLH) is widely used as a carrier protein for conjugation to haptens and other antigens to make them more immunogenic for the purpose of antibody production. Because of its large mass and complexity, KLH elicits a stronger immune response than other carrier proteins. Because the protein is derived from a mollusk, it is phylogenetically distant from mammalian species and less likely to produce antibodies that will cross-react with typical target samples in assays.
More Product Data
• Carrier protein activation and conjugation data for immunogen preparation
Related Products
Imject™ Maleimide-Activated mcKLH Spin Kit
Imject™ Maleimide PEGylated mcKLH
Imject™ Maleimide Conjugation Buffer
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity10 x 10 mg
Product LineImject
Product TypeImject Maleimide Activated Mariculture KLH
Unit SizeEach
Contents & Storage
Upon receipt store product at 4°CC. Product is shipped with an ice pack.