Halt™ Protease Inhibitor Cocktail (100X)
Thermo Scientific™

Halt™ Protease Inhibitor Cocktail (100X)

Thermo Scientific Halt Protease Inhibitor Cocktail (100X) are ready-to-use concentrated stock solutions of protease inhibitors for addition to samples to자세히 알아보기
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호수량
7843024 x 100 μL
784295 mL
7843810 mL
877861 mL
카탈로그 번호 78430
제품 가격(KRW)
308,000
온라인 행사
Ends: 30-Sep-2026
342,000
할인액 34,000 (10%)
Each
수량:
24 x 100 μL
대량 주문 또는 맞춤형 요청
제품 가격(KRW)
308,000
온라인 행사
Ends: 30-Sep-2026
342,000
할인액 34,000 (10%)
Each
Thermo Scientific Halt Protease Inhibitor Cocktail (100X) are ready-to-use concentrated stock solutions of protease inhibitors for addition to samples to prevent proteolytic degradation during cell lysis and protein extraction.

Features of Halt Protease Inhibitor Cocktail, (100X):

• Convenient—the refrigerator-stable, 100X liquid format is more effective and easier to use than individual inhibitors or other formats; just pipette exactly the amount you need
• No proprietary ingredients—fully disclosed formulation contains optimized concentrations of six broad-spectrum protease inhibitors AEBSF, aprotinin, bestatin, E-64, leupeptin and pepstatin A stabilized in high-quality dimethylsulfoxide (DMSO)
• Multiple package sizes—choose among several package sizes, including single-use microtubes that keep protease inhibitors fresh and reduce the risk of reagent contamination
• Regular formulation—includes a separate vial of EDTA solution for optional metalloprotease inhibition
• Lysis buffer compatible—use with Thermo Scientific Pierce Cell Lysis Buffers or nearly any other commercial or homemade detergent-based lysis reagent. Add 10 μL of concentrated cocktail per 1 mL of lysis buffer to ensure complete protection of the resulting protein extract.

Applications:
• Protection of intact, active cellular proteins from degradation by endogenous proteases
• Inhibition of protease activity from lysates produced by a variety of methods including Thermo Scientific Pierce Cell Lysis Reagents, other commercially formulated cell or tissue lysis products, sonication, French press, blender homogenization or repetitive freeze/thaw cycling
• Screening of extracts for proteolytic activity
• The study of proteolysis in the regulation of cellular processes

Proteases make up a large category of enzymes, including endopeptidases and exopeptidases, which catalyze the hydrolytic breakdown of proteins into peptides or amino acids in a wide variety of cell and tissue types. Proteases are regulated and compartmentalized in living cells, but cell lysis releases and mixes them with other extracted proteins. Their activity can thus lead to the loss of a large number of valuable proteins, adversely affecting downstream applications. For this reason, it is common practice to spike cell lysis buffers or cell extracts with a cocktails of compounds known to inactivate or inhibit proteases. Halt Protease Inhibitor Cocktails effectively inhibit serine-proteases, cysteine-proteases, aspartic acid-proteases and aminopeptidases that are typically present in cellular lysate samples. Regular and EDTA-free formulations provide for inhibition of metalloproteases and compatibility with 2D electrophoresis, respectively.

Related Products
Halt™ Protease Inhibitor Single-Use Cocktail, EDTA-free (100X)

본 제품은 냉장/냉동제품으로 반송된 제품은 전량 폐기 처리 되오니 주문 전 상세 내용 다시 한번 확인 부탁드립니다.
For Research Use Only. Not for use in diagnostic procedures.
사양
버퍼Lysis Buffer
억제제Protease Inhibitor
수량24 x 100 μL
시약 유형Protease Inhibitor
충분240 mL Sample
형태Liquid
제품라인Halt
제품 유형Protease Inhibitor Cocktail
Unit SizeEach
구성 및 보관
Upon receipt store at 4°C

자주 묻는 질문(FAQ)

What are the compositions of the Halt Protease Inhibitor Cocktail, EDTA-Free (100X) (Cat. No. 87785) and Halt Protease Inhibitor Cocktail (100X) (Cat. No. 87786)?

Both formulations are the same and have the following composition:

What is the shelf life of Halt Protease Inhibitor Cocktail (100X)?

When stored as recommended at 4 degrees C, Halt Protease Inhibitor Cocktail (100X) is guaranteed stable for 1 year from the date of receipt.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

Can I use just part of the Pierce Protease Inhibitor XL Capsules, EDTA-Free and save part of the capsule for later use?

We do not recommend “splitting” the capsule into smaller portions. Researchers using less than 500 mL should choose a different format (liquid or tablet) of protease inhibitors.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

What formats of protease inhibitors do you offer?

We offer standalone protease inhibitors (AEBSF, Aprotinin, PMSF, EDTA, etc.), as well as cocktails or combinations of protease inhibitors in liquid, tablet, and larger capsule formats. These cocktails of protease inhibitors can also be purchased in combination with a cocktail of phosphatase inhibitors, in both liquid and tablet form, for even greater convenience.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

What protease inhibtors are present in the Halt Protease Inhibitor Single-Use Cocktail?

The inhibitors included in the Halt Protease Inhibitor Single-Use Cocktail (Cat. No. 78430) are: AEBSF, Aprotinin, Bestatin, E64, Leupeptin, Pepstatin A, and EDTA.

인용 및 참조 문헌 (5)

인용 및 참조 문헌
Abstract
Modification of histidine repeat proteins by inorganic polyphosphate.
Authors:Neville N,Lehotsky K,Yang Z,Klupt KA,Denoncourt A,Downey M,Jia Z
Journal:Cell reports
PubMed ID:37660293
Inorganic polyphosphate (polyP) is a linear polymer of orthophosphate that is present in nearly all organisms studied to date. A remarkable function of polyP involves its attachment to lysine residues via non-enzymatic post-translational modification (PTM), which is presumed to be covalent. Here, we show that proteins containing tracts of consecutive ... More
Deletion of the prorenin receptor in the ureteric bud in mice inhibits Dot1/H3K79 pathway.
Authors:Song R,Yosypiv IV
Journal:Pediatric research
PubMed ID:38287105
BACKGROUND: The prorenin receptor (PRR) plays a critical role in ureteric bud (UB) branching morphogenesis. DOT1 Like (DOT1L), a histone methyltransferase specific for Histone 3 lysine 79 (H3K79), is important for differentiation of the UB-derived renal collecting duct cells. In this study, we tested whether DOT1L/H3 dimethyl K79 (H3m2K79) are ... More
Metformin inhibits digestive proteases and impairs protein digestion in mice.
Authors:Kelly CJ,Verdegaal AA,Anderson BW,Shaw WL,Bencivenga-Barry NA,Folta-Stogniew E,Goodman AL
Journal:The Journal of biological chemistry
PubMed ID:37863262
Metformin is among the most prescribed medications worldwide and the first-line therapy for type 2 diabetes. However, gastrointestinal side effects are common and can be dose limiting. The total daily metformin dose frequently reaches several grams, and poor absorption results in high intestinal drug concentrations. Here, we report that metformin ... More
β-Cell-specific ablation of sirtuin 4 does not affect nutrient-stimulated insulin secretion in mice.
Authors:Huynh FK,Peterson BS,Anderson KA,Lin Z,Coakley AJ,Llaguno FMS,Nguyen TN,Campbell JE,Stephens SB,Newgard CB,Hirschey MD
Journal:American journal of physiology. Endocrinology and metabolism
PubMed ID:32865009
Sirtuins are a family of proteins that regulate biological processes such as cellular stress and aging by removing posttranslational modifications (PTMs). We recently identified several novel PTMs that can be removed by sirtuin 4 (SIRT4), which is found in mitochondria. We showed that mice with a global loss of SIRT4 ... More
Glucagon-like peptide 1 inhibits cell apoptosis and improves glucose responsiveness of freshly isolated human islets.
Authors:Farilla L, Bulotta A, Hirshberg B, Li Calzi S, Khoury N, Noushmehr H, Bertolotto C, Di Mario U, Harlan DM, Perfetti R
Journal:Endocrinology
PubMed ID:12960095
The peptide hormone, glucagon-like peptide 1 (GLP-1), has been shown to increase glucose-dependent insulin secretion, enhance insulin gene transcription, expand islet cell mass, and inhibit beta-cell apoptosis in animal models of diabetes. The aim of the present study was to evaluate whether GLP-1 could improve function and inhibit apoptosis in ... More