Fiberlite™ F14-6 x 250y Fixed Angle Rotor, for Sorvall™ RC-6 Plus, Sorvall™ Evolution RC - FAQs
Fiberlite™ F14-6 x 250y Fixed Angle Rotor, for Sorvall™ RC-6 Plus, Sorvall™ Evolution RC - FAQs
View additional product information for Fiberlite™ F14-6 x 250y Fixed Angle Rotor, for Sorvall™ RC-6 Plus, Sorvall™ Evolution RC - FAQs (78500)
This product does not have any FAQs available.