Thermo Sequenase Cycle Sequencing Kit - FAQs
Thermo Sequenase Cycle Sequencing Kit - FAQs
View additional product information for Thermo Sequenase Cycle Sequencing Kit - FAQs (785001KT)
1 product FAQs found
I'm looking for the user guide for the Thermo Sequenase Cycle Sequencing Kit (Cat. No. 785001KT). Can you please provide the link?
Here is the link to the Thermo Sequenase Cycle Sequencing Kit user guide: https://www.affymetrix.com/support/technical/usb/proto/78500a.pdf
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Applications Support Center.