Orion™ 8030cX Silica Analyzer
Thermo Scientific™

Orion™ 8030cX Silica Analyzer

Confidently perform silica analysis with the Thermo Scientific™ Orion™ 8030cX Silica Analyzer. Designed to provide accurate, effortless water analysis, the Orion 8030cX specializes in online measurement of reactive silica.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoTipoCantidad
8030REXKit de reactivo de sílice1 unidad
7070FLTPrefiltro de muestras1 unidad
803002Solución estándar de sílice20 μg/L
803020Solución estándar de sílice200 μg/L
803030Solución estándar de sílice1000 μg/L
803040Solución estándar de sílice4000 μg/L
8030CLSolución de limpieza1 unidad
8030CXAnalizador de sílice1 unidad
8030MKKit de mantenimiento1 unidad
8010PCMTarjeta Profibus1 unidad
Número de catálogo 8030REX
Precio (MXN)
-
Tipo:
Kit de reactivo de sílice
Cantidad:
1 unidad
Confidently perform silica analysis with the Thermo Scientific™ Orion™ 8030cX Silica Analyzer. Designed to provide accurate, effortless water analysis, the Orion 8030cX specializes in online measurement of reactive silica. Powered by advanced colorimetric technology, the Orion 8030cX delivers premium precision, greater productivity and enhanced durability. Supporting ultrapure and process water applications, the Orion 8030cX conveniently delivers precise, reliable measurement you can depend on. Unleash the possibilities and conquer the freedom of fast, confident decision making.
Confident Results
  • Low Detection Limit (0.5 ppb)
  • High Accuracy (≤1 ppb or 1% for range 0-500 ppb)
  • Patented Fluid Delivery & Quantification System
  • Reduced Phosphate Interferences

 Low Cost Ownership
  • 100 Days of Continuous Run Time (w/ 2L reagent and 15-minute measurement frequency)
  • Low Reagent Consumption (200 μL per measurement)
  • Separate Chemical Waste Collection (decreased disposal expenses)

 Easy to Use
  • Automated Calibration
  • Automated Validation & Verification
  • Color Touchscreen with Intuitive Interface

 Maximum Uptime
  • Automated Cleaning
  • Scheduled Measurements
  • Alert Notification System
  • Alarm System w/ Troubleshooting Guide
  • IP65 Certified (splash/ dust proof)
Markets
Silica monitoring for ultrapure water, demineralized water, deionized water, boiler water and steam condensate in:
  • Power Generation
  • Oil & Gas
  • Chemical Production
  • Semiconductor Manufacturing
  • Pulp & Paper


Applications

Monitoring and controlling silica levels in ultrapure and industrial process water to:
  • Prevent buildup on turbine blades, heat exchangers, boilers and more
  • Reduce costly and time-consuming cleaning procedures
  • Identify reverse osmosis (RO) breakdown and saturation of ion exchange resins
  • Maximize thermal efficiencies of steam passageways
  • Safeguard the health of critical plant assets by reducing potential for turbine vibration
Especificaciones
DescripciónThe Thermo Scientific™ Orion™ 8030cX Silica Reagent Kit includes three (3) 2L bottles of our custom reagent formulation, including reagents 1, 2 and 3.
Para utilizar con (equipo)Orion 8030cX Silica Analyzer
Cantidad1 unidad
TipoKit de reactivo de sílice
Unit SizePack of