Forma™ 8600 Series -86°C Ultra-Low Temperature Chest Freezer, 3 cu. ft. (84.9L) capacity, 120V/60Hz, 10.5A - FAQs
Forma™ 8600 Series -86°C Ultra-Low Temperature Chest Freezer, 3 cu. ft. (84.9L) capacity, 120V/60Hz, 10.5A - FAQs
View additional product information for Forma™ 8600 Series -86°C Ultra-Low Temperature Chest Freezers - FAQs (803CA)