CD22 PerCP-eFluor 710, ASR, eBioscience™ - FAQs
CD22 PerCP-eFluor 710, ASR, eBioscience™ - FAQs
View additional product information for CD22 PerCP-eFluor 710, ASR, eBioscience™ - FAQs (8046-4229-120)
1 product FAQs found
Can the eFluor Organic Dyes be frozen?
As with other organic fluorochromes, we do not recommend the eFluor Organic Dyes be frozen.