CD9 APC-eFluor™ 780, ASR, eBioscience™ - FAQs
CD9 APC-eFluor™ 780, ASR, eBioscience™ - FAQs
View additional product information for CD9 APC-eFluor™ 780, ASR, eBioscience™ - FAQs (8047-0098-120)
1 product FAQs found
Can the eFluor Organic Dyes be frozen?
As with other organic fluorochromes, we do not recommend the eFluor Organic Dyes be frozen.