CD10 APC-eFluor 780, ASR, eBioscience™ - FAQs
CD10 APC-eFluor 780, ASR, eBioscience™ - FAQs
View additional product information for CD10 APC-eFluor 780, ASR, eBioscience™ - FAQs (8047-0108-120)
1 product FAQs found
Can the eFluor Organic Dyes be frozen?
As with other organic fluorochromes, we do not recommend the eFluor Organic Dyes be frozen.