Ig Kappa eFluor 450, ASR, eBioscience™ - FAQs
Ig Kappa eFluor 450, ASR, eBioscience™ - FAQs
View additional product information for Ig Kappa eFluor 450, ASR, eBioscience™ - FAQs (8048-9970-120)
1 product FAQs found
Can the eFluor Organic Dyes be frozen?
As with other organic fluorochromes, we do not recommend the eFluor Organic Dyes be frozen.