Orion™ Storage Sleeve and Base
Thermo Scientific™

Orion™ Storage Sleeve and Base

Protect electrodes from breakage when not in use with the Thermo Scientific™ Orion™ Electrode Storage Sleeve and Base.
Have Questions?
Catalog NumberType
810017Storage Sleeve, Base
Catalog number 810017
Price (USD)
67.77
Online exclusive
75.30
Save 7.53
Each
Type:
Storage Sleeve, Base
Price (USD)
67.77
Online exclusive
75.30
Save 7.53
Each
Protect electrodes from breakage when not in use with the Thermo Scientific™ Orion™ Electrode Storage Sleeve and Base.

The Thermo Scientific Orion Storage Sleeve and Base is for use with pH electrodes that are 120mm or less in length that have a diameter of 12mm. The sleeve protects your electrode from breakage when not in use and will not tip, due to the weighted removable base. As an additional convenience, the pH bulb stays conditioned while in storage so that it is ready to use when removed from the sleeve.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionOrion Storage Sleeve and Base
For Use With (Equipment)pH Electrode
TypeStorage Sleeve, Base
Unit SizeEach