CaptureSelect™ AVB Sepharose HP Leakage ELISA
Thermo Scientific™

CaptureSelect™ AVB Sepharose HP Leakage ELISA

Thermo Scientific™ CaptureSelect™ AVB Sepharose HP Leakage ELISA is designed to detect possible leached ligand of adeno-associated viruses (AAV) purified using AVB Sepharose HP affinity resin (Cytiva).
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
8102808011 Assay
81028081010 Assays
Catalog number 810280801
Price (USD)
1,250.00
Each
Quantity:
1 Assay
Price (USD)
1,250.00
Each

CaptureSelect AVB Sepharose HP Leakage ELISA is designed to detect possible leached ligand of adeno-associated viruses (AAV) purified using AVB Sepharose HP affinity resin (Cytiva). Even when covalently attached, small amounts of the AVB Sepharose HP affinity ligand can leach off of the chromatography support and co-elute with the virus particles.

This ELISA provides typical sensitivity and accurate quantitation of possible leached ligand greater than or equal to 1 ng/mL. The CaptureSelect AVB Sepharose HP Leakage ELISA can be used as a tool to aid in optimal purification process development and in routine quality control of in-process streams as well as final product.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
AssayELISA
Detection Limit1 ng/mL
For Use With (Equipment)Microplate Reader
FormatIndividual components
Product LineCaptureSelect
Product TypeLigand Leakage ELISA
Quantity1 Assay
Shipping ConditionDry Ice
TargetAVB Sepharose HP Affinity Ligand
Detection MethodAbsorbance at 450 nm
Immunoassay Kit FormatSandwich ELISA Kit
Sample TypeLigand Leachate
Target SpeciesNot Applicable
Unit SizeEach
Contents & Storage
Store at -20°C.