CaptureSelect™ VIII Select Leakage ELISA
Thermo Scientific™

CaptureSelect™ VIII Select Leakage ELISA

Thermo Scientific™ CaptureSelect™ VIIISelect Leakage ELISA is designed to detect possible leached ligand of B domain-depleted recombinant Factor VIII purified using VIIISelect affinity resin (Cytiva).
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
8102860011 Assay
81028601010 Assays
Catalog number 810286001
Price (USD)
1,250.00
Each
Quantity:
1 Assay
Price (USD)
1,250.00
Each

CaptureSelect VIIISelect Leakage ELISA is designed to detect possible leached ligand of B domain-depleted recombinant Factor VIII purified using VIIISelect affinity resin (Cytiva). Even when covalently attached, small amounts of the VIIISelect affinity ligand can leach off of the chromatography support and co-elute with human Factor VIII.

This ELISA provides typical sensitivity and accurate quantitation of possible leached ligand greater than or equal to 1 ng/mL. The CaptureSelect VIIISelect Leakage ELISA can be used as a tool to aid in optimal purification process development and in routine quality control of in-process streams as well as final product.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
AssayELISA
Detection LimitDetection of ≤1 ng/mL VIII Select Affinity Ligand
For Use With (Equipment)Microplate Reader
FormatIndividual components
Product LineCaptureSelect
Product TypeLigand Leakage ELISA
Quantity1 Assay
Shipping ConditionDry Ice
TargetVIIISelect Affinity Ligand
Detection MethodAbsorbance at 450 nm
Immunoassay Kit FormatSandwich ELISA Kit
Sample TypeLigand Leachate
Target SpeciesNot Applicable
Unit SizeEach
Contents & Storage
Store at -20°C.