CaptureSelect™ Alpha-1 Antitrypsin Select Leakage ELISA
Thermo Scientific™

CaptureSelect™ Alpha-1 Antitrypsin Select Leakage ELISA

Thermo Scientific™ CaptureSelect™ Alpha-1 AntitrypsinSelect Leakage ELISA is designed to detect possible leached ligand of human Alpha-1 antitrypsin (AAT) purified using AAT Select affinity resin (Cytiva).
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
81028701010 Assays
8102870011 Assay
Catalog number 810287010
Price (USD)
11,230.00
Each
Quantity:
10 Assays
Price (USD)
11,230.00
Each

CaptureSelect Alpha-1 AntitrypsinSelect Leakage ELISA is designed to detect possible leached ligand of human Alpha-1 antitrypsin (AAT) purified using AAT Select affinity resin (Cytiva). Even when covalently attached, small amounts of the Alpha-1 Antitrypsin Select affinity ligand can leach off of the chromatography support and co-elute with the purified product.

This ELISA provides typical sensitivity and accurate quantitation of possible leached ligand greater than or equal to 1 ng/mL. The CaptureSelect Alpha-1 Antitrypsin Select Leakage ELISA can be used as a tool to aid in optimal purification process development and in routine quality control of in-process streams as well as final product.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
AssayELISA
Detection Limit1 ng/mL
For Use With (Equipment)Microplate Reader
FormatIndividual components
Product LineCaptureSelect
Product TypeLigand Leakage ELISA
Quantity10 Assays
Shipping ConditionDry Ice
TargetAlpha-1 AntitrypsinSelect Affinity Ligand
Detection MethodAbsorbance at 450 nm
Immunoassay Kit FormatSandwich ELISA Kit
Sample TypeLigand Leachate
Target SpeciesNot Applicable
Unit SizeEach
Contents & Storage
Store at -20°C.