CaptureSelect™ VII Select Leakage ELISA
Thermo Scientific™

CaptureSelect™ VII Select Leakage ELISA

Thermo Scientific™ CaptureSelect™ VIISelect Leakage ELISA is designed to detect possible leached ligand of human Factor VII purified using VIISelect affinity resin (Cytiva).
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
8102990011 Assay
81029901010 Assays
Catalog number 810299001
Price (USD)
1,250.00
Each
Quantity:
1 Assay
Price (USD)
1,250.00
Each

CaptureSelect VIISelect Leakage ELISA is designed to detect possible leached ligand of human Factor VII purified using VIISelect affinity resin (Cytiva). Even when covalently attached, small amounts of the VIISelect affinity ligand can leach off of the chromatography support and co-elute with the purified product.

This ELISA provides typical sensitivity and accurate quantitation of possible leached ligand greater than or equal to 1 ng/mL. The CaptureSelect VIISelect Leakage ELISA can be used as a tool to aid in optimal purification process development and in routine quality control of in-process streams as well as final product.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
AssayELISA
Detection Limit1 ng/mL
For Use With (Equipment)Microplate Reader
FormatIndividual components
Product LineCaptureSelect
Product TypeLigand Leakage ELISA
Quantity1 Assay
Shipping ConditionDry Ice
TargetVIISelect Affinity Ligand
Detection MethodAbsorbance at 450 nm
Immunoassay Kit FormatSandwich ELISA Kit
Sample TypeLigand Leachate
Target SpeciesNot Applicable
Unit SizeEach
Contents & Storage
Store at -20°C.