CaptureSelect™ C-tagXL Ligand Leakage ELISA
Thermo Scientific™

CaptureSelect™ C-tagXL Ligand Leakage ELISA

Thermo Scientific™ CaptureSelect™ C-tagXL Ligand Leakage ELISA is designed to detect possible leached ligand of C-terminal E-P-E-A tagged target proteins purified using CaptureSelect C-tagXL Affinity Matrix.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCantidad
8103072011 ensayo
81030721010 ensayos
Número de catálogo 810307201
Precio (MXN)
-
Cantidad:
1 ensayo

CaptureSelect C-tagXL Ligand Leakage ELISA is designed to detect possible leached ligand of C-terminal E-P-E-A tagged target proteins purified using CaptureSelect C-tagXL Affinity Matrix. The ligand, a 13 kD recombinant protein, is covalently coupled to give high chemical stability and low leakage. Even when covalently attached, small amounts of the affinity ligand can leach off of the chromatography support and co-elute with the protein.

This ELISA provides typical sensitivity and accurate quantitation of possible leached ligand greater than or equal to 1 ng/mL. The CaptureSelect C-tagXL Ligand Leakage ELISA can be used as a tool to aid in optimal purification process development and in routine quality control of in-process streams as well as final product.

For Research Use Only. Not for use in diagnostic procedures.
Especificaciones
Límite de detección1 ng/mL
FormatoKit
Tipo de ligandoCaptureSelect C-tagXL Affinity Ligand
Línea de productosCaptureSelect
Tipo de productoLigand Leakage ELISA
Cantidad1 ensayo
Condiciones de envíoDry Ice
Método de detecciónAbsorbance at 450 nm
Formato del kit de inmunoensayoSandwich ELISA Kit
Tipo de muestraLigand Leachate
Unit SizeEach
Contenido y almacenamiento
Store in freezer (-5°C to -30°C).