CaptureSelect™ GCSFSelect Leakage ELISA
Thermo Scientific™

CaptureSelect™ GCSFSelect Leakage ELISA

Thermo Scientific™ CaptureSelect™ GCSFSelect Leakage ELISA is designed to detect possible leached ligand of human GCSF purified using GCSFSelect affinity resin (Cytiva).
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
81031301010 Assays
8103130011 Assay
Catalog number 810313010
Price (USD)
11,230.00
Each
Quantity:
10 Assays
Price (USD)
11,230.00
Each

CaptureSelect GCSFSelect Leakage ELISA is designed to detect possible leached ligand of human GCSF purified using GCSFSelect affinity resin (Cytiva). Even when covalently attached, small amounts of the IgSelect affinity ligand can leach off of the chromatography support and co-elute with the antibody.

This ELISA provides typical sensitivity and accurate quantitation of possible leached ligand greater than or equal to 1 ng/mL. The CaptureSelect GCSFSelect Leakage ELISA can be used as a tool to aid in optimal purification process development and in routine quality control of in-process streams as well as final product.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
AssayELISA
Detection Limit1 ng/mL
For Use With (Equipment)Microplate Reader
FormatIndividual components
Product LineCaptureSelect
Product TypeLigand Leakage ELISA
Quantity10 Assays
Shipping ConditionDry Ice
TargetGCSFSelect Affinity Ligand
Detection MethodAbsorbance at 450 nm
Immunoassay Kit FormatSandwich ELISA Kit
Sample TypeLigand Leachate
Target SpeciesNot Applicable
Unit SizeEach
Contents & Storage
Store at -20°C.