CaptureSelect™ FSH Ligand Leakage ELISA
Thermo Scientific™

CaptureSelect™ FSH Ligand Leakage ELISA

Thermo Scientific™ CaptureSelect™ FSH Ligand Leakage ELISA is designed to detect possible leached ligand of follicle-stimulating hormone (FSH) purified using CaptureSelect FSH Affinity Matrix.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
8103180011 Assay
81031801010 Assays
Catalog number 810318001
Price (USD)
1,250.00
Each
Quantity:
1 Assay
Price (USD)
1,250.00
Each

CaptureSelect FSH Ligand Leakage ELISA is designed to detect possible leached ligand of follicle-stimulating hormone (FSH) purified using CaptureSelect FSH Affinity Matrix. The ligand, a 14 kD recombinant protein, is coupled by multipoint attachment through stable amide bonds to give high chemical stability and low leakage. Even when covalently attached, small amounts of the affinity ligand can leach off of the chromatography support and co-elute with the protein.

This ELISA provides typical sensitivity and accurate quantitation of possible leached ligand greater than or equal to 1 ng/mL. The CaptureSelect FSH Ligand Leakage ELISA can be used as a tool to aid in optimal purification process development and in routine quality control of in-process streams as well as final product.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
AssayELISA
Detection Limit1 ng/mL
For Use With (Equipment)Microplate Reader
FormatIndividual components
Product LineCaptureSelect
Product TypeLigand Leakage ELISA
Quantity1 Assay
Shipping ConditionDry Ice
TargetCaptureSelect FSH Affinity Ligand
Detection MethodAbsorbance at 450 nm
Immunoassay Kit FormatSandwich ELISA Kit
Sample TypeLigand Leachate
Target SpeciesNot Applicable
Unit SizeEach
Contents & Storage
Store at -20°C.