CaptureSelect™ AAV9 Ligand Leakage ELISA
Thermo Scientific™

CaptureSelect™ AAV9 Ligand Leakage ELISA

Thermo Scientific™ CaptureSelect™ AAV9 Ligand Leakage ELISA is designed to detect possible leached ligand of adeno-associated viruses (AAV) purified using POROS CaptureSelect AAV9 Affinity Resin.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
81033301010 Assays
8103330011 Assay
Catalog number 810333010
Price (USD)
11,230.00
Each
Quantity:
10 Assays
Price (USD)
11,230.00
Each

CaptureSelect AAV9 Ligand Leakage ELISA is designed to detect possible leached ligand of adeno-associated viruses (AAV) purified using POROS CaptureSelect AAV9 Affinity Resin. The ligand, a 13 kD recombinant protein, is coupled by multipoint attachment through stable amide bonds to give high chemical stability and low leakage. Even when covalently attached, small amounts of the affinity ligand can leach off of the chromatography support and co-elute with the purified AAV.

This ELISA provides typical sensitivity and accurate quantitation of possible leached ligand greater than or equal to 1 ng/mL. The CaptureSelect AAV9 Ligand Leakage ELISA can be used as a tool to aid in optimal purification process development and in routine quality control of in-process streams as well as final product.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
AssayELISA
Detection Limit1 ng/mL
For Use With (Equipment)Microplate Reader
FormatIndividual components
Product LineCaptureSelect
Product TypeLigand Leakage ELISA
Quantity10 Assays
Shipping ConditionDry Ice
TargetCaptureSelect AAV9 Affinity Ligand
Detection MethodAbsorbance at 450 nm
Immunoassay Kit FormatSandwich ELISA Kit
Sample TypeLigand Leachate
Target SpeciesNot Applicable
Unit SizeEach
Contents & Storage
Store at -20°C.