CaptureSelect™ AAV8 Ligand Leakage ELISA
Thermo Scientific™
CaptureSelect™ AAV8 Ligand Leakage ELISA
Thermo Scientific™ CaptureSelect™ AAV8 Ligand Leakage ELISA is designed to detect possible leached ligand of adeno-associated viruses (AAV) purified using POROS CaptureSelect AAV8 Affinity Resin.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
81033801010 Assays
8103380011 Assay
Catalog number 810338010
Price (USD)
11,230.00
Each
Quantity:
10 Assays
Price (USD)
11,230.00
Each
CaptureSelect AAV8 Ligand Leakage ELISA is designed to detect possible leached ligand of adeno-associated viruses (AAV) purified using POROS CaptureSelect AAV8 Affinity Resin. In this resin the ligand, a 13 kD recombinant protein, is coupled by multipoint attachment through stable amide bonds to give high chemical stability and low leakage. Even when covalently attached, small amounts of the affinity ligand can leach off of the chromatography support and co-elute with the purified AAV.
This ELISA provides typical sensitivity and accurate quantitation of possible leached ligand greater than or equal to 1 ng/mL. The CaptureSelect AAV8 Ligand Leakage ELISA can be used as a tool to aid in optimal purification process development and in routine quality control of in-process streams as well as final product.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)Microplate Reader
Product LineCaptureSelect
Quantity10 Assays
Detection MethodColorimetric
Unit SizeEach
Contents & Storage
3 x 1 mL vials, store at -5°C to -30°C