CaptureSelect™ Factor X Ligand Leakage ELISA
Thermo Scientific™

CaptureSelect™ Factor X Ligand Leakage ELISA

Thermo Scientific™ CaptureSelect™ Factor X Ligand Leakage ELISA is designed to detect possible leached ligand of Factor X purified using CaptureSelect Factor X Affinity Matrix.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
81037021010 assay x 96-well plate
8103702011 assay x 96-well plate
Catalog number 810370210
Price (USD)
11,240.00
Each
Quantity:
10 assay x 96-well plate
Price (USD)
11,240.00
Each

The Factor X affinity ligand is a 13-kD recombinant protein expressed in yeast and covalently coupled to a chromatography matrix to give high chemical stability and low leakage. During the chromatography process small amounts of the affinity ligand can leach off of the chromatography support and co-elute with the protein. This ELISA provides typical sensitivity and accurate quantitation of possible leached ligand greater than or equal to 1 ng/mL.

The CaptureSelect Factor X Ligand Leakage ELISA is designed to minimize interference and to provide accurate quantitation even in the presence of the protein of interest. It can be used as a tool to aid in optimal purification process development and in routine quality control of in-process streams as well as final product.

Specifications
Detection LimitDetection of ≤1 ng/mL Factor X Affinity Ligand
FormatIndividual components
Product LineCaptureSelect
Product TypeLigand Leakage ELISA
Quantity10 assay x 96-well plate
Shipping ConditionDry Ice
Sufficient For10 x 96-well plates
TargetCaptureSelect Factor X Affinity Ligand
Detection MethodAbsorbance at 450 nm
Immunoassay Kit FormatSandwich ELISA Kit
Sample TypeLigand Leachate
Target SpeciesNot Applicable
Unit SizeEach
Contents & Storage
Store at -20°C.